missing translation for 'onlineSavingsMsg'
Learn More

CD142 Mouse anti-Human, Brilliant Violet 711, Clone: HTF-1, BD Optibuild™

Código de producto. 15818709
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15818709 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15818709 Proveedor BD Biosciences N.º de proveedor 744007

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The HTF-1 monoclonal antibody specifically binds to CD142. CD142 is a 45-47 kDa, single chain, type I transmembrane glycoprotein also known as Tissue Factor (TF). CD142 has been referred in the literature as coagulation Factor III or thromboplastin and it is expressed on activated endothelial cells and lipopolysaccharide (LPS)-stimulated monocytes/macrophages. TF associates with factor VIIa to form a complex and acts as an enzyme that initiates the blood coagulation cascade. It is known as the major initiator of clotting in normal hemostasis. CD142 can be induced by various inflammatory mediators in monocytes and vascular endothelial cells. This antibody is useful in inflammation, thrombosis and hemostasis research.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD142
Clasificación Monoclonal
Clon HTF-1
Concentración 0.2mg/mL
Conjugado Brilliant Violet 711
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen Tissue factor, TF, TFA, Coagulation factor III, F3, Thromboplastin
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.