missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD14 Monoclonal Antibody (TuK4), FITC

Descripción
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Especificaciones
Especificaciones
| Antígeno | CD14 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | TuK4 |
| Concentración | 0.20mg/mL |
| Conjugado | FITC |
| Descripción | CD14 Monoclonal Antibody, 0.5mL, 0.20mg/mL Concentration, TuK4 Clone, CD14 Antigen, Human CD14 Immunogen, Mouse Host, Human Species Reactivity, FITC Conjugate, IgG2a Isotype, PBS with 4mg/mL BSA Storage Buffer, Used For Flow Cytometry |
| Dilución | Assay-dependent |
| Formulación | PBS with 4mg/mL BSA and 0.1% sodium azide |
| génica | Cd14 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?