Learn More
CD138 (Syndecan-1) Mouse, anti-Human, PE, Clone: MI15, STEMCELL Technologies
Código de producto. 17148341
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17148341 100 Tests 100 tests
17158341 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17148341 Proveedor Stemcell Technologies N.º de proveedor 60003PE
Mouse Monoclonal Antibody
  • The MI15 antibody reacts with an extracellular epitope on CD138 (Syndecan-1), an ∼85 - 95 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells), as well as on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
  • CD138 expression is used as a diagnostic marker for several types of tumors
  • CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
  • Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
  • The MI15 antibody recognizes a different epitope than that of the clone DL-101 anti-CD138 antibody but blocks binding of clone B-B4
Especificaciones
Antígeno CD138 (Syndecan-1)
Aplicaciones Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon MI15
Conjugado PE
Especie del huésped Mouse
Cantidad 100 Tests
Primario o secundario Primary
Especies diana Cynomolgus, Human, Rhesus Monkey
Isotype IgG1 κ
Tipo de célula B Cells, Plasma