missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138 Monoclonal Antibody (MI15), Invitrogen™
Descripción
MA5-12400 targets CD138 in IHC (P), FACS, IP, and WB applications and shows reactivity with Human and Rat samples. The MA5-12400 immunogen is u-266 cells.
CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
Especificaciones
Especificaciones
| Antígeno | CD138 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | MI15 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | tissue culture supernatant with 0.09% sodium azide |
| génica | Sdc1 |
| N.º de referencia del gen | P18827, P26260 |
| Alias de gen | AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?