missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CD137 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. p-7244918
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17834413 50 μg 50 microgramos
17884673 100 μg 100 microgramos
17804083 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17834413

Marca: PeproTech 500P167G50UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

AA Sequence of recombinant protein: MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human 4-1BB Receptor. Anti-Human 4-1BB Receptor-specific antibody was purified by affinity chromatography employing an immobilized Human 4-1BB Receptor matrix. Sandwich ELISA: To detect Human 4-1BB Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human 4-1BB Receptor (500-P167GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human 4-1BB Receptor. Western Blot: To detect Human 4-1BB Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human 4-1BB Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD137
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica TNFRSF9
N.º de referencia del gen Q07011
Alias de gen 4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Símbolos de los genes TNFRSF9
Especie del huésped Goat
Inmunógeno E.coli-derived Recombinant Human 4-1BB Receptor.
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3604
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.