missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD137 Monoclonal Antibody (BBK-2)
Descripción
MA5-13739 targets CD137 in FACS and IF applications and shows reactivity with Human samples. The MA5-13739 immunogen is ectodomain of human 4-1BB recombinant protein.
CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
Especificaciones
Especificaciones
| Antígeno | CD137 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | BBK-2 |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 |
| génica | TNFRSF9 |
| N.º de referencia del gen | Q07011 |
| Alias de gen | 4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?