missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD130 (GP130) Recombinant Rabbit Monoclonal Antibody (BLR114H)

Descripción
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Especificaciones
Especificaciones
| Antígeno | CD130 (GP130) |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | BLR114H |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | BBS with 0.09% sodium azide; pH 8.2 |
| génica | IL6ST |
| N.º de referencia del gen | P40189 |
| Alias de gen | 5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha |
| Mostrar más |
Título del producto
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?