missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127/IL7R Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17626-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD127 (Interleukin-7, IL-7) is a glycoprotein involved in the regulation of lymphopoiesis. The response of cells to CD127 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is an alpha/gamma chain heterodimer. CD127 consists of an alpha chain and a gamma chain. The gamma (c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. The biological effects of IL7 are mediated by the binding of IL7 to the specific cell surface receptor. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.Especificaciones
| CD127/IL7R | |
| Polyclonal | |
| Unconjugated | |
| IL7R | |
| CD127, CDW127, IL 7 receptor subunit alpha, IL 7R alpha, IL 7R subunit alpha, IL 7RA, IL7R, IL7RA, ILRA, interleukin 7 receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16197, 3575 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16871, P16872 | |
| IL7R | |
| CD127/IL7R Fusion Protein Ag11844 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido