missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD127 (Interleukin-7, IL-7) is a glycoprotein involved in the regulation of lymphopoiesis. The response of cells to CD127 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is an alpha/gamma chain heterodimer. CD127 consists of an alpha chain and a gamma chain. The gamma (c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. The biological effects of IL7 are mediated by the binding of IL7 to the specific cell surface receptor. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.
Especificaciones
Especificaciones
| Antígeno | CD127/IL7R |
| Aplicaciones | Flow Cytometry, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.15 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | IL7R |
| N.º de referencia del gen | P16871, P16872 |
| Alias de gen | CD127, CDW127, IL 7 receptor subunit alpha, IL 7R alpha, IL 7R subunit alpha, IL 7RA, IL7R, IL7RA, ILRA, interleukin 7 receptor |
| Símbolos de los genes | IL7R |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?