Learn More
CD11c/Integrin alpha X Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16887214 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16887214 20 μL 20 microlitros
16877214 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16887214 Proveedor Proteintech N.º de proveedor 173421AP20UL
Rabbit Polyclonal Antibody
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
Especificaciones
Antígeno CD11c/Integrin alpha X
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ITGAX
N.º de referencia del gen P20702
Alias de gen CD11C, Integrin alpha X, ITGAX, Leu M5, SLEB6
Símbolos de los genes ITGAX
Especie del huésped Rabbit
Inmunógeno CD11c/Integrin alpha X Fusion Protein Ag11380
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3687
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG