missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Hamster, anti-Mouse, Pacific Blue, Clone: N418, STEMCELL Technologies
Descripción
- The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor
- Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis
- In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780)
Especificaciones
Especificaciones
| Antígeno | CD11c |
| Aplicaciones | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | N418 |
| Conjugado | Pacific Blue |
| Especie del huésped | Hamster |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Isotype | IgG |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?