missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 (c-Kit) Rat, anti-Mouse, APC, Clone: 2B8, STEMCELL Technologies
Descripción
- The 2B8 antibody reacts with CD117 (c-Kit), an ∼145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells
- CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain
- Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells
- The 2B8 antibody does not block binding of c-Kit ligand
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Hematopoietic Progenitor Cell Isolation Kit (Catalog #19856)
Especificaciones
Especificaciones
| Antígeno | CD117 (c-Kit) |
| Aplicaciones | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | 2B8 |
| Conjugado | APC |
| Especie del huésped | Rat |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Isotype | IgG2b κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?