missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD109 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA582535
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: GPVEILTTVT ESVTGISRNV STNVFFKQHD YIIEFFDYTT VLKPSLNFTA TVKVTRADGN QLTLEERRNN VVITVTQRNY TEYWSGSNSG NQKMEAVQKI NYTVPQSGTF KIEFPILEDS SELQLKAYFL GSKSSMA Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 61%.
CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
Especificaciones
| CD109 | |
| Polyclonal | |
| Unconjugated | |
| CD109 | |
| 150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 135228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6YHK3 | |
| CD109 | |
| Recombinant protein corresponding to Human CD109. Recombinant protein control fragment (Product #RP-89854). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido