missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 (Endoglin) Mouse, anti-Human, Alexa Fluor 488, Clone: 43A3, STEMCELL Technologies
Descripción
- The 43A3 antibody reacts with CD105 (endoglin), an ∼180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ∼90 kDa type I transmembrane subunits
- CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromal fibroblasts
- Its expression is also observed in some types of tumors, and levels are up-regulated on the endothelium during angiogenesis
- CD105 has important roles in angiogenesis, cardiovascular development, and vascular remodeling, and the protein serves a regulatory role in cytoskeletal reorganization by modulating the sites of focal adhesion and cellular migration
- This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Especificaciones
Especificaciones
| Antígeno | CD105 (Endoglin) |
| Aplicaciones | Immunoprecipitation, Western Blot, Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 43A3 |
| Conjugado | Alexa Fluor 488 |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Human, Mouse |
| Isotype | IgG1 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?