Learn More
CD103 Hamster anti-Mouse, PE, Clone: 2E7, Proteintech
Hamster Monoclonal Antibody
Marca: Proteintech PE-65047-100UG
Descripción
The CD103 antibody from Proteintech is a hamster monoclonal antibody generated with c57bl/6j mouse intestinal intraepithelial lymphocytes (iel) of mouse CD103. This antibody recognizes mouse antigen. The CD103 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Especificaciones
| CD103 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Hamster | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| 2E7 | |
| PE | |
| 100 μg | |
| 16407 | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.