Learn More
CCR8 (CD198) Mouse anti-Human , APC, Clone: 433H, BD Biosciences™
Código de producto. 17227694
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17227694 25 Tests 25 tests
17217694 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17227694 Proveedor BD Biosciences N.º de proveedor 566898
Mouse Monoclonal Antibody
CCR8 (CD198) Monoclonal antibody specifically detects CCR8 (CD198) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CCR8 (CD198)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 433H
Conjugado APC
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Alias de gen CCR8; CKR-L1; CKRL1; CMKBR8; CMKBRL2; CY6; GPR-CY6; GPRCY6; MGC129966; MGC1
Especie del huésped Mouse
Inmunógeno Human CCR8 Transfected Cell Line
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed.
Isotype IgG2a κ