missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV786, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566758
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Especificaciones
| CCR7 (CD197) | |
| Monoclonal | |
| Brilliant Violet 786 | |
| BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
| Human CCR7 Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 2-L1-A | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido