missing translation for 'onlineSavingsMsg'
Learn More

CCR7 (CD197) Mouse anti-Human , BV711, Clone: 2-L1-A, BD Biosciences™

Código de producto. 17207584
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
Código de producto. Cantidad unitSize
17207584 25 Tests 25 tests
17297574 100 Tests 100 tests
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17207584

Marca: BD Biosciences 566753

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CCR7 (CD197)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 2-L1-A
Conjugado Brilliant Violet 711
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Alias de gen BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
Especie del huésped Mouse
Inmunógeno Human CCR7 Transfected Cell Line
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV711 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV711 were removed.
Isotype IgG1 κ
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.