missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , APC, Clone: 2-L1-A, BD Biosciences™
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antígeno | CCR7 (CD197) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 2-L1-A |
| Conjugado | APC |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Alias de gen | BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor |
| Especie del huésped | Mouse |
| Inmunógeno | Human CCR7 Transfected Cell Line |
| Cantidad | 25 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?