missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR10 Recombinant Rabbit Monoclonal Antibody (HL2852)

Descripción
CCR10 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines. CCR10 is the receptor for CCL27; CCR10-CCL27 interactions are involved in T cell-mediated skin inflammation.
Especificaciones
Especificaciones
| Antígeno | CCR10 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | HL2852 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | CCR10 |
| N.º de referencia del gen | P46092 |
| Alias de gen | CC chemokine receptor 10; C-C chemokine receptor type 10; C-C CKR-10; C-C motif chemokine receptor 10; CC-CKR-10; CCR10; CCR-10; chemokine (C-C motif) receptor 10; chemokine C-C receptor 9; Cmkbr9; G protein-coupled receptor 2; Gpr2; G-protein coupled receptor 2; MGC151420; RP23-281C18.3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?