missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CCL1 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. p-7245164
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17884083 50 μg 50 microgramos
17894033 100 μg 100 microgramos
17874323 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17884083

Marca: PeproTech 500P11050UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human I-309 (CCL1). Anti-Human I-309 (CCL1)-specific antibody was purified by affinity chromatography employing an immobilized Human I-309 (CCL1) matrix. Sandwich ELISA: To detect Human I-309 (CCL1) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human I-309 (CCL1) (500-P110Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human I-309 (CCL1). Western Blot: To detect Human I-309 (CCL1) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human I-309 (CCL1) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CCL1
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica CCL1
N.º de referencia del gen P22362
Alias de gen BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3
Símbolos de los genes CCL1
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human I-309 (CCL1).
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6346
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.