missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCKBR Polyclonal Antibody

Descripción
PA3-201 detects CCK2 in human samples. PA3-201 has successfully been used in Western blot and immunocytochemical procedures. The PA3-201 immunogen is a synthetic peptide corresponding to the C-terminal amino acids R(428)ALPDEDPPTPSIASLSRLSYTTISTLGPG(447) of human CCK2.
CCKBR is a G-protein coupled receptor for gastrin and cholecystokinin (CCK), regulatory peptides of the brain and gastrointestinal tract. This protein is a type B gastrin receptor, which has a high affinity for both sulfated and nonsulfated CCK analogs and is found principally in the central nervous system and the gastrointestinal tract. A misspliced transcript variant including an intron has been observed in cells from colorectal and pancreatic tumors. CCKBR has been reported in brain, colon, pancreas, stomach, thyroid, and various cancers.
Especificaciones
Especificaciones
| Antígeno | CCKBR |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Whole serum with 0.02% sodium azide |
| génica | CCKBR |
| N.º de referencia del gen | P32239 |
| Alias de gen | CCK(B) receptor; CCK2 receptor; Cck2r; CCK2-R; CCKB; CCK-B; Cckb receptor; CCK-B receptor; Cckbr; CCK-BR; CCKB-R; CCKR-2; CCKRB; cholecystokinin B receptor; cholecystokinin-2 receptor; Cholecystokinin-B; Cholrec; GASR; gastrin receptor; gastrin/cholecystokinin type B receptor; HGNC:1571 |
| Símbolos de los genes | CCKBR |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?