missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCF642, MedChemExpress
Description
CCF642 is a potent protein disulfide isomerases (PDI) inhibitor with an IC50 of 2.9 μM. CCF642 causes acute endoplasmic reticulum (ER) stress in multiple myeloma cells accompanied by apoptosis-inducing calcium release. CCF642 has broad anti-multiple myeloma activity.
Specifications
Specifications
| Nombre del producto químico o material | CCF642 |
| CAS | 346640-08-2 |
| Color | Naranja |
| Notas de grado de pureza | Research |
| Fórmula molecular | C15H10N2O4S3 |
| Cantidad | 10 mg |
| Información de solubilidad | DMSO : ≥ 30 mg/mL (79.27 mM) |
| SMILES | O=C1N(C2=CC=C(OC)C=C2)C(S/C1=C/C3=CC=C([N+]([O-])=O)S3)=S |
| Peso molecular (g/mol) | 378.45 |
| Formula Weight (peso de la fórmula) | 378.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?