Descripción
CCDC93 is involved in copper-dependent ATP7A trafficking between the trans-Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes and is dependent on its interaction with WASHC2C.
Especificaciones
Especificaciones
| Antígeno | CCDC93 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.19 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | CCDC93 |
| N.º de referencia del gen | Q567U6, Q5BJT7, Q7TQK5 |
| Alias de gen | CCDC93 |
| Símbolos de los genes | CCDC93 |
| Mostrar más |