missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC93 Monoclonal Antibody (OTI6E11), TrueMAB™, OriGene
Descripción
CCDC93 Monoclonal antibody specifically detects CCDC93 in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Especificaciones
Especificaciones
| Antígeno | CCDC93 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI6E11 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | JUN |
| N.º de referencia del gen | P05412, P05627, P17325 |
| Alias de gen | 4633402D15Rik; 9230102M16Rik; CCDC93; coiled-coil domain containing 93; coiled-coil domain-containing protein 93 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?