missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC171 Polyclonal Antibody

Descripción
Immunogen sequence: ASTRIMTLEK EMTSHRSHIA ALKSELHTAC LRENASLQSI GSRDHSNLSI PSRAPLPADT TGIGDFLPLK AELDTTYTFL KETFINTVPH ALTSSHSSPV TMSANANRPT QI Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 26%.
Chromosome 9 consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias. There are two isoforms of C9orf93 that are produced as a result of alternative splicing events.
Specifications
Specifications
| Antígeno | CCDC171 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | Lot-Specific |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | CCDC171 |
| N.º de referencia del gen | Q6TFL3 |
| Alias de gen | bA536D16.1; bA778P13.1; C9orf93; CCDC171; coiled-coil domain containing 171; coiled-coil domain-containing protein 171; myosin tail domain containing protein |
| Símbolos de los genes | CCDC171 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?