missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC155 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA546337
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: VIHETSEETE FPSEAPAGGQ RNFQGEPAHP EEGRKEPSMW LTRREEEEDA Sequence homology: Cow: 77%; Human: 100%.
As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex, involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Required for telomere attachment to nuclear envelope in the prophase of meiosis and for rapid telomere prophase movements implicating a SUN1/2:KASH5 LINC complex in which SUN1 and SUN2 seem to act at least partial redundantly. Required for homolog pairing during meiotic prophase in spermatocytes and probably oocytes. Essential for male and female gametogenesis. Recruits cytoplasmic dynein to telomere attachment sites at the nuclear envelope in spermatocytes. In oocytes is involved in meiotic resumption and spindle formation. [UniProt].
Especificaciones
| CCDC155 | |
| Polyclonal | |
| Unconjugated | |
| CCDC155 | |
| CCDC155; coiled-coil domain containing 155; coiled-coil domain-containing protein 155; KASH domain containing 5; KASH domain-containing protein 5; KASH5; protein KASH5; Unknown (protein for MGC:134376) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 147872 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N6L0 | |
| CCDC155 | |
| Synthetic peptide directed towards the C-terminal of human CCDC155 (aa 404-453). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido