Learn More
Invitrogen™ CCDC92 Polyclonal Antibody

Description
Immunogen sequence: MTSPHFSSYD EGPLDVSMAA TNLENQLHSA QKNLLFLQRE HASTLKGLHS EIRRLQQHCT DLTYELTVKS SEQTGDGTSK SSELKKRCEE LEAQLKVKEN ENAELLKELE QKNAMITVLE NTIKEREKKY LEELKAKSHK LTLLSSELEQ RASTIAYLTS QLHAAKKKLM SSSGTSDASP SGSPVLASYK PAPPKDKLPE TPRRRMKKSL SAPLHPEFEE VYRFGAESRK LLLREPVDAM PDPTPFLLAR ESAEVHLIKE RPLVIPPIAS DRSGEQHSPA REKPHKAHVG VAHRIHHATP PQAQPEVKTL AVDQVNGGKV VRKHSGTDRT V.
Specifications
Specifications
| Antígeno | CCDC92 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.57 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | Ccdc92 |
| N.º de referencia del gen | Q53HC0 |
| Alias de gen | AW050178; Ccdc92; coiled-coil domain containing 92; coiled-coil domain-containing protein 92; D5Bwg0834e; limkain beta 2; limkain beta-2 |
| Símbolos de los genes | Ccdc92 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.