missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCAR1 Polyclonal Antibody

Beschreibung
This antibody is predicted to react with bovine, canine, human and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
IFI-44 (interferon-induced protein 44), also known as p44 or MTAP44 (microtubule-associated protein 44), is a 444 amino acid protein that localizes to the cytoplasm and, upon induction by IFN-betas, aggregates to form microtubular structures. Human IFI-44 shares 97% sequence similarity with its chimp counterpart, suggesting a conserved role between species. The gene encoding IFI-44 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
Spezifikation
Spezifikation
| Antígeno | CCAR1 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| génica | CCAR1 |
| N.º de referencia del gen | Q8CH18, Q8IX12 |
| Alias de gen | 2610511G16Rik; 9430036H15Rik; Carp1; CARP-1; Ccar1; Cell cycle and apoptosis regulatory protein 1; cell division cycle and apoptosis regulator 1; cell division cycle and apoptosis regulator protein 1; Death inducer with SAP domain; DIS; MGC44628; OTTHUMP00000019709; RGD1560358; RP11-437A18.1 |
| Símbolos de los genes | CCAR1 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?