Descripción
The heteroteramer with HSD17B8 has NADH-dependent 3-ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria. The homotetramer may act as NADPH-dependent quinone reductase. Has broad substrate specificity and reduces 9,10-phethrenequinone, 1,4-benzoquinone and various other o-quinones and p-quinones (in vitro).
Especificaciones
Especificaciones
| Antígeno | CBR4 |
| Aplicaciones | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | CBR4 |
| N.º de referencia del gen | Q7TS56, Q8N4T8, Q91VT4 |
| Alias de gen | carbonyl reductase 4, CBR4, Quinone reductase CBR4, SDR45C1 |
| Símbolos de los genes | Cbr4 |
| Mostrar más |