missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA539079
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
Especificaciones
| CBR3 | |
| Polyclonal | |
| Unconjugated | |
| CBR3 | |
| SDR21C2, hCBR3, NADPH-dependent carbonyl reductase 3, carbonyl reductase (NADPH) 3, carbonyl reductase [NADPH] 3, short chain dehydrogenase/reductase family 21C, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109857, 304078, 874 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75828, Q8K354 | |
| CBR3 | |
| A synthetic peptide derived from the internal region of human CBR3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido