missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CBFB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CBFB. This antibody recognizes human, rat antigen. The CBFB antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.
Specifications
Specifications
| Antígeno | CBFB |
| Aplicaciones | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Clasificación | Monoclonal |
| Clon | 1D7F2 |
| Concentración | 1000 μg/mL |
| Conjugado | Unconjugated |
| Símbolos de los genes | PEBP2B |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Protein |
| Método de purificación | Protein G purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?