missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaV2.1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15634565
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15634565 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15634565

Marca: Invitrogen™ PA550634

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CACNA1A protein.

Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas, the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1A subunit, which is predominantly expressed in neuronal tissue. Mutations in this gene are associated with 2 neurologic disorders, familial hemiplegic migraine and episodic ataxia 2. This gene also exhibits polymorphic variation due to n-repeats. Multiple transcript variants encoding different isoforms have been found for this gene. In one set of transcript variants, the n-repeats occur in the 3 UTR, and are not associated with any disease. But in another set of variants, an insertion extends the coding region to include the n-repeats which encode a polyglutamine tract. Expansion of the n-repeats from the normal 4-16 to 21-28 in the coding region is associated with spinocerebellar ataxia 6.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CaV2.1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
génica CACNA1A
N.º de referencia del gen O00555
Alias de gen alpha1A; APCA; BccA1; BI; brain calcium channel 1; brain calcium channel I; brain class A; Caca1a; Cach4; CACN3; CACNA1A; Cacnl1a4; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; calcium voltage-gated channel subunit alpha1 A; Cav2.1; Ccha1a; EA2; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; FHM; HPCA; la; leaner; MHP; MHP1; nmf352; Rat brain class A; rbA-1; RBA-I; rkr; rocker; SCA6; tg; tottering; voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1
Símbolos de los genes CACNA1A
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to a region derived from internal residues of human calcium channel, voltage-dependent, P/Q type, alpha 1A subunit.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 773
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.