missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaV2.1 Polyclonal Antibody
Descripción
For Western Blot, this antibody has non-specific bands at 42 kDa. This antibody is predicted to react with rabbit, cow and human based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas, the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1A subunit, which is predominantly expressed in neuronal tissue. Mutations in this gene are associated with 2 neurologic disorders, familial hemiplegic migraine and episodic ataxia 2. This gene also exhibits polymorphic variation due to n-repeats. Multiple transcript variants encoding different isoforms have been found for this gene. In one set of transcript variants, the n-repeats occur in the 3 UTR, and are not associated with any disease. But in another set of variants, an insertion extends the coding region to include the n-repeats which encode a polyglutamine tract. Expansion of the n-repeats from the normal 4-16 to 21-28 in the coding region is associated with spinocerebellar ataxia 6.
Especificaciones
Especificaciones
| Antígeno | CaV2.1 |
| Aplicaciones | Immunohistochemistry (Frozen), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 98.98% PBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| génica | CACNA1A |
| N.º de referencia del gen | P54282, P97445 |
| Alias de gen | alpha1A; APCA; BccA1; BI; brain calcium channel 1; brain calcium channel I; brain class A; Caca1a; Cach4; CACN3; CACNA1A; Cacnl1a4; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; calcium voltage-gated channel subunit alpha1 A; Cav2.1; Ccha1a; EA2; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; FHM; HPCA; la; leaner; MHP; MHP1; nmf352; Rat brain class A; rbA-1; RBA-I; rkr; rocker; SCA6; tg; tottering; voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1 |
| Símbolos de los genes | CACNA1A |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?