missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 8 p18 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 16384315 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16384315 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16384315

Marca: Invitrogen™ PA595047

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: A549 whole cell, HELA whole cell. IHC: Human Tonsil Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Caspase 8 (CASP8) is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases play a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a pro-domain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. Caspase 8 is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of Caspase 8 suggests that it may interact with Fas-interacting protein FADD. Caspase 8 was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the pro-form of the Caspase 8 protein. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby committing the cell to undergo apoptosis. In addition, Caspase 8 also reacts with Jurkat cells and Tonsil. Overexpression of Caspase 8 induces apoptosis, which can be blocked by inhibitors specific for the ICE family. Many alternatively spliced transcript variants encoding different isoforms have been described for Caspase 8, however, not all variants have had their full-length sequences determined.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Caspase 8 p18
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 500 μg/mL
Conjugado Unconjugated
Formulación PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
génica Casp8
N.º de referencia del gen Q14790
Alias de gen ALPS2B; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; Casp8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; caspase-8; Caspase-8 subunit p10; Caspase-8 subunit p18; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MORT1-associated ced-3 homolog
Símbolos de los genes Casp8
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to a sequence at the N-terminus of human Caspase-8 (p18) (257-277aa NNHNFAKAREKVPKLHSIRDR).
Método de purificación Affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 841
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.