Learn More
CARS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Novus Biologicals NBP2-32597
 Ver más versiones de este producto
Código de producto. 18751654
  • ONLINE EXCLUSIVE PRICE
    589.00€
    556.50€ / 0.10 ml
    Ahorro 32.50€
    5% Descuento
Descripción
CARS2 Polyclonal specifically detects CARS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Western Blot
0.1 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Certificados
Promociones
Promociones
For Research Use Only