Learn More
Melford Carrageenan Kappa
Código de producto. p-200087273
Click to view available options
Cantidad:
100 g
250 g
Tamaño de la unidad:
100 g
250 g
Código de producto. 30018162
Marca: Melford C41070100.0
Kappa carrageenan binds water to form strong, rigid gels in the presence of potassium salts.
As the level of potassium is increased, the gel structure becomes tightly aggregated and may result in syneresis (moisture on the gel surface).
Especificaciones
Cantidad 100 g