Learn More
R&D Systems™ CardioTACS In Situ Apoptosis Detection Kit
Descripción
Includes
Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 1.5mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2; Other reagents
Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform
Complete kits contain TACS BlueLabel™, Nuclear Fast Red counterstain and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in cardiac tissues
Study fixed cells, embedded tissues and froze
Especificaciones
Especificaciones
| Cantidad | 30 pruebas |
| Contenido y almacenamiento | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |