Learn More
R&D Systems™ CardioTACS In Situ Apoptosis Detection Kit
Código de producto. 18411860
Change view
Click to view available options
Cantidad:
30 pruebas
Tamaño de la unidad:
30 tests
18411860 30 pruebas 30 tests
1 options
Código de producto. 18411860 Proveedor R&D Systems™ N.º de proveedor 482730K
Includes
Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 1.5mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2; Other reagents
Provides all the reagents for the detection of DNA fragmentation in cardiac cells and tissue sections along with a cation that specifically enhances the signal in apoptotic cardiac cells
Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform
Complete kits contain TACS BlueLabel™, Nuclear Fast Red counterstain and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in cardiac tissues
Study fixed cells, embedded tissues and froze
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 30 pruebas
Contenido y almacenamiento The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.