Learn More
Cardiac Troponin T Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16814724 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16814724 20 μL 20 microlitros
16804724 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16814724 Proveedor Proteintech N.º de proveedor 155131AP20UL
Rabbit Polyclonal Antibody
Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.
Especificaciones
Antígeno Cardiac Troponin T
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Concentración 0.19 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TNNT2
N.º de referencia del gen P45379, P50752, P50753
Alias de gen Cardiac Troponin T, CMH2, cTnT, TNNT2, TnTC
Símbolos de los genes TNNT2
Especie del huésped Rabbit
Inmunógeno Cardiac Troponin T Fusion Protein Ag7716
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21956, 24837, 7139
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG