missing translation for 'onlineSavingsMsg'
Learn More

Cardiac Troponin I Mouse anti-Human, Mouse, Rat, Clone: 1D5D6, Proteintech

Código de producto. 16878195 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16878195 20 μL 20 microlitros
16868195 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16878195 Proveedor Proteintech N.º de proveedor 663761IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This antibody is specific to cTnI, but not cross-react with skeletal TnI.

Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).

Specifications

Antígeno Cardiac Troponin I
Aplicaciones Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Clasificación Monoclonal
Clon 1D5D6
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TNNI3
N.º de referencia del gen P19429, P23693, P48787
Alias de gen Cardiac troponin I, CMD2A, CMH7, cTnI, RCM1, TNNC1, TNNI3, troponin I type 3 (cardiac), Troponin I, cardiac muscle
Símbolos de los genes TNNI3
Especie del huésped Mouse
Inmunógeno Cardiac Troponin I Fusion Protein Ag16758
Método de purificación Protein G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21954, 29248, 7137
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.