missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase III Polyclonal Antibody
GREENER_CHOICE

Product Code. 16367524
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
Product Code. Cantidad unitSize
16367524 100 μL 100 microlitros
1 options
This item is not returnable. View return policy

Product Code. 16367524

Brand: Invitrogen™ PA580392

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Carbonic Anhydrase III/CA3.

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Carbonic Anhydrase III
Aplicaciones ELISA, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica Ca3
N.º de referencia del gen P07451
Alias de gen BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Símbolos de los genes Ca3
Especie del huésped Rabbit
Inmunógeno Recombinant Human Carbonic Anhydrase III/CA3 protein (Met1-Lys260).
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 761
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.