Learn More
CAPRIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15112-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Especificaciones
| CAPRIN1 | |
| Polyclonal | |
| Unconjugated | |
| CAPRIN1 | |
| caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362173, 4076, 53872 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14444, Q5M9G3, Q60865 | |
| Caprin1 | |
| CAPRIN1 Fusion Protein Ag7099 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |