missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAP1 Recombinant Rabbit Monoclonal Antibody (23GB3020), Invitrogen™
Descripción
Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.
Especificaciones
Especificaciones
| Antígeno | CAP1 |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB3020 |
| Concentración | 0.6 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | CAP1 |
| N.º de referencia del gen | Q01518 |
| Alias de gen | adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?