missing translation for 'onlineSavingsMsg'
Learn More

CANX Mouse anti-Human, Clone: CANX/1541, Abnova™

Código de producto. 16043296
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16043296 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16043296 Supplier Abnova Supplier No. MAB15046.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human CANX.

  • High specificity
  • High reactivity

Specifications

Antígeno CANX
Aplicaciones Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Clasificación Monoclonal
Clon CANX/1541
Conjugado Unconjugated
Descripción Mouse monoclonal antibody raised against partial recombinant human CANX.
Dilución Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined b
Formulación In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
N.º de referencia del gen P27824
Alias de gen CNX/FLJ26570/IP90/P90
Símbolos de los genes CANX
Especie del huésped Mouse
Inmunógeno Recombinant protein corresponding to N-terminus of human CANX.
Método de purificación Protein A/G purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 821
Especies diana Human
Contenido y almacenamiento Store at 4°C.
Formulario Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.