missing translation for 'onlineSavingsMsg'
Learn More

Canine coronavirus Goat anti-Virus, Polyclonal, Abnova™

Product Code. 16058277
Change view
Click to view available options
Cantidad:
1 mg
Unit Size:
1 miligramo
Product Code. Cantidad unitSize
16058277 1 mg 1 miligramo
1 options
This item is not returnable. View return policy

Product Code. 16058277

Brand: Abnova PAB29835.1mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

Goat polyclonal antibody raised against canine coronavirus.

  • High specificity
  • High reactivity

Specifications

Antígeno Canine coronavirus
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Goat polyclonal antibody raised against canine coronavirus.
Dilución ELISA Western Blot The optimal working dilution should be determined by the end user.
Formulación In PBS, pH 7.2 (0.09% sodium azide).
Especie del huésped Goat
Inmunógeno Native purified canine coronavirus nucleocapsid protein.
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento Store at 4°C. Do not freeze.
Formulario Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.