Learn More
CAMSAP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16887874 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16887874 150 μL 150 microlitros
16897874 20 μL 20 microlitros
2 options
Código de producto. 16887874
Marca: Proteintech 178801AP150UL
Rabbit Polyclonal Antibody
The CAMSAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CAMSAP2. This antibody recognizes human, mouse, rat antigen. The CAMSAP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Especificaciones
Antígeno CAMSAP2
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Polyclonal
Concentración 393 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 23271
Especies diana Human, Rat, Mouse
Isotype IgG