Learn More
DiscoverX cAMP Hunter™ eXpress OPRL1 CHO-K1 GPCR Assay
Descripción
The cAMP Hunter™ OPRL1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. orphanin FQ) based activation of OPRL1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype OPRL1 , a naturally Gi/Go coupled GPCR. Upon activation of OPRL1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | CHO-K1 |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | Orphanin FQ |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | OPRL1 |
| Línea de células | CHO-K1 |
| Objetivo | OPRL1 |