Learn More
DiscoverX cAMP Hunter™ CHO-K1 OPRD1 Gi Cell Line
Descripción
The cAMP Hunter OPRD1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. DADLE, DPDPE) based activation of OPRD1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype OPRD1, a naturally Gi/Go coupled GPCR. Upon activation of OPRD1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Especificaciones
Especificaciones
| Entrada del ensayo | Intracellular cAMP level |
| Línea de células | CHO-K1 |
| Para utilizar con (aplicación) | Cell Lines |
| Agonista primario | [D-Ala2, D-Leu5]-Enkephalin (DADLE) |
| Cantidad | 2 viales |
| Entrada diana | OPRD1 |
| Número de acceso | NM_000911 |