Learn More
DiscoverX cAMP Hunter™ CHO-K1 GHRHR Gs Cell Line
Descripción
The cAMP Hunter GHRHR Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GRF) based activation of GHRHR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GHRHR , a naturally Gs coupled GPCR. Upon activation of GHRHR , the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Especificaciones
Especificaciones
| Entrada del ensayo | Intracellular cAMP level |
| Línea de células | CHO-K1 |
| Para utilizar con (aplicación) | Cell Lines |
| Agonista primario | Growth Hormone Releasing Factor, GRF (1-29) |
| Cantidad | 2 viales |
| Entrada diana | GHRHR |
| Número de acceso | NM_000823.2 |