missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (010), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA543469
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human CAMKI/CAMK1
CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Especificaciones
CaMKI | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q14012 | |
CAMK1 | |
Recombinant Human CAMKI/CAMK1 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
010 | |
Unconjugated | |
CAMK1 | |
AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 | |
Rabbit | |
Protein A | |
RUO | |
8536 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (010), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido